Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quickfit
®
air condenser
Products
Manufacturer
Length (mm)
Surface Area cm²
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z303089
Quickfit
®
air condenser,
L 200 mm, joint: ST/NS 14/23
Expand
View Pricing
Z303119
Quickfit
®
air condenser,
L 300 mm, joint: ST/NS 24/29
Expand
View Pricing
Z303127
Quickfit
®
air condenser,
L 500 mm, joint: ST/NS 24/29
Expand
View Pricing
Z303097
Quickfit
®
air condenser,
L 150 mm, joint: ST/NS 19/26
Expand
View Pricing