Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Q-Gard
®
Purification Cartridge
Products
Product Category
polishing cartridges (12)
Brand
Milli-Q (12)
Compatibility
Quality Segment
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
QGARDT1X1
Q-Gard
®
Purification Cartridge,
For Milli-Q
®
Reference and Advantage A10 systems connected to Elix
®
/ RO / distilled water feed
Expand
View Pricing
QGARDT2X1
Q-Gard
®
Purification Cartridge,
For Milli-Q
®
Advantage A10 / Reference systems connected to DI water feed
Expand
View Pricing
QGARD00R1
Q-Gard
®
Purification Cartridge,
For Milli-Q
®
Element (2001-2009) / Academic / Biocel / Element / Gradient / Synthesis systems connected to Elix
®
/ RO / distilled water feed
Expand
View Pricing
QGARD00D2
Q-Gard
®
Purification Cartridge,
For Milli-Q
®
Academic / Biocel / Element / Gradient / Synthesis water purification systems connected to DI water feed
Expand
View Pricing
QGARDTXL04
Q-Gard
®
Purification Cartridge,
For use with Milli-Q
®
CLX/HX/HX SD/HR 7000 series systems, Produces high resistivity water from purification media.
Expand
View Pricing
CP2ALLREST
Q-Gard
®
Purification Cartridge,
Polishing cartridge for AFS
®
8/16 and AFS
®
8D/16D systems
Expand
View Pricing
QGARDTL01
Q-Gard
®
Purification Cartridge,
For Elix
®
Gulfstream / Large and RiOs
™
Large systems connected to a pure water feed source
Expand
View Pricing
QGARD00D3
Q-Gard
®
Purification Cartridge,
For Milli-Q
®
Element / Academic / Biocel / Element / Gradient / Synthesis systems connected to DI water feed containing high silt
Expand
View Pricing
QGARDT3X1
Q-Gard
®
Purification Cartridge,
For Milli-Q
®
Advantage A10 / Reference systems connected to DI water feed containing high silt
Expand
View Pricing
QGARDTBX1
Q-Gard
®
Purification Cartridge,
For Milli-Q
®
Advantage A10/Reference systems, for dedicated removal of boron traces from water
Expand
View Pricing
QGARD00B1
Q-Gard
®
Purification Cartridge,
For dedicated removal of boron traces from water dispensed by Milli-Q
®
Element / Academic / Biocel / Element / Gradient / Synthesis systems
Expand
View Pricing
CP2ALLRES
Q-Gard
®
Purification Cartridge,
Polishing cartridge for AFS
®
8/16 and AFS
®
8D/16D systems
Expand
View Pricing