
Pyridinium dichromate

Synonym(s):
PDC
Empirical Formula (Hill Notation):
C10H10N2 · H2Cr2O7
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing