Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Test tubes, without rim, medium wall
Products
Product Category
tubes (10)
Manufacturer
Capacity (mL)
Diameter (mm)
OD (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
SLW1622/04M
Pyrex
®
Test tubes, without rim, medium wall,
6 mL
Expand
View Pricing
SLW1622/09M
Pyrex
®
Test tubes, without rim, medium wall,
14 mL
Expand
View Pricing
SLW1622/20M
Pyrex
®
Test tubes, without rim, medium wall,
50 mL
Expand
View Pricing
SLW1622/12M
Pyrex
®
Test tubes, without rim, medium wall,
22 mL
Expand
View Pricing
SLW1622/15M
Pyrex
®
Test tubes, without rim, medium wall,
34 mL
Expand
View Pricing
SLW1622/06M
Pyrex
®
Test tubes, without rim, medium wall,
8 mL
Expand
View Pricing
SLW1622/22M
Pyrex
®
Test tubes, without rim, medium wall,
66 mL
Expand
View Pricing
SLW1622/10M
Pyrex
®
Test tubes, without rim, medium wall,
18 mL
Expand
View Pricing
SLW1622/14M
Pyrex
®
Test tubes, without rim, medium wall,
23 mL
Expand
View Pricing
SLW1622/16M
Pyrex
®
Test tubes, without rim, medium wall,
28 mL
Expand
View Pricing