Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Screwcaps for Media-lab bottle
Products
Product Category
caps (7)
Manufacturer
Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
SLW4506/45D
Pyrex
®
Screwcaps for Media-lab bottle,
light blue, thread 45 mm
Expand
View Pricing
SLW4506/45G
Pyrex
®
Screwcaps for Media-lab bottle,
gray, thread 45 mm
Expand
View Pricing
SLW4506/45F
Pyrex
®
Screwcaps for Media-lab bottle,
yellow, thread 45 mm
Expand
View Pricing
SLW4506/45E
Pyrex
®
Screwcaps for Media-lab bottle,
green, thread 45 mm
Expand
View Pricing
SLW4506/80D
Pyrex
®
Screwcaps for Media-lab bottle,
blue, thread 80 mm
Expand
View Pricing
SLW4506/25D
Pyrex
®
Screwcaps for Media-lab bottle,
light blue, thread 25 mm
Expand
View Pricing
SLW4506/32D
Pyrex
®
Screwcaps for Media-lab bottle,
light blue, thread 32 mm
Expand
View Pricing