Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Griffin beakers, low form, with printed trace code
Products
Product Category
beakers (15)
Manufacturer
Diameter (mm)
Height (mm)
Color
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
SLW1000/04D
Pyrex
®
Griffin beakers, low form, with printed trace code,
100 mL
Expand
View Pricing
SLW1000/10D
Pyrex
®
Griffin beakers, low form, with printed trace code,
250 mL
Expand
View Pricing
SLW1000/02M
Pyrex
®
Griffin beakers, low form, with printed trace code,
50 mL
Expand
View Pricing
SLW1000/34D
Pyrex
®
Griffin beakers, low form, with printed trace code,
5000 mL
Expand
View Pricing
SLW1000/05M
Pyrex
®
Griffin beakers, low form, with printed trace code,
10 mL
Expand
View Pricing
SLW1000/03M
Pyrex
®
Griffin beakers, low form, with printed trace code,
5 mL
Expand
View Pricing
SLW1000/22D
Pyrex
®
Griffin beakers, low form, with printed trace code,
1000 mL
Expand
View Pricing
SLW1000/11M
Pyrex
®
Griffin beakers, low form, with printed trace code,
25 mL
Expand
View Pricing
SLW1000/18D
Pyrex
®
Griffin beakers, low form, with printed trace code,
600 mL
Expand
View Pricing
SLW1000/14D
Pyrex
®
Griffin beakers, low form, with printed trace code,
400 mL
Expand
View Pricing
SLW1000/30D
Pyrex
®
Griffin beakers, low form, with printed trace code,
3000 mL
Expand
View Pricing
SLW1000/06D
Pyrex
®
Griffin beakers, low form, with printed trace code,
150 mL
Expand
View Pricing
SLW1000/38D
Pyrex
®
Griffin beakers, low form, with printed trace code,
10000 mL
Expand
View Pricing
SLW1000/28D
Pyrex
®
Griffin beakers, low form, with printed trace code,
2000 mL
Expand
View Pricing
SLW1000/20D
Pyrex
®
Griffin beakers, low form, with printed trace code,
800 mL
Expand
View Pricing