Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Griffin beakers, heavy duty, graduated
Products
Product Category
beakers (9)
Manufacturer
Diameter (mm)
Height (mm)
Color
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
SLW1003-2L
Pyrex
®
Griffin beakers, heavy duty, graduated,
2000 mL
Expand
View Pricing
SLW1003-1L
Pyrex
®
Griffin beakers, heavy duty, graduated,
1000 mL
Expand
View Pricing
SLW1003-5L
Pyrex
®
Griffin beakers, heavy duty, graduated,
5000 mL
Expand
View Pricing
SLW1003-250
Pyrex
®
Griffin beakers, heavy duty, graduated,
250 mL
Expand
View Pricing
SLW1003-3L
Pyrex
®
Griffin beakers, heavy duty, graduated,
3000 mL
Expand
View Pricing
SLW1003-600
Pyrex
®
Griffin beakers, heavy duty, graduated,
600 mL
Expand
View Pricing
SLW1003-150
Pyrex
®
Griffin beakers, heavy duty, graduated,
150 mL
Expand
View Pricing
SLW1003-4L
Pyrex
®
Griffin beakers, heavy duty, graduated,
4000 mL
Expand
View Pricing
SLW1003-400
Pyrex
®
Griffin beakers, heavy duty, graduated,
400 mL
Expand
View Pricing