Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
disposable culture tubes, screw cap style with white marking spot
Products
Product Category
tubes (6)
Color
Manufacturer
Capacity (mL)
OD (mm)
Cap
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS9944713
Pyrex
®
disposable culture tubes, screw cap style with white marking spot,
capacity 7.5 mL
Expand
View Pricing
CLS9944716
Pyrex
®
disposable culture tubes, screw cap style with white marking spot,
capacity 15.5 mL
Expand
View Pricing
CLS99447161
Pyrex
®
disposable culture tubes, screw cap style with white marking spot,
capacity 11.5 mL
Expand
View Pricing
CLS9944720X
Pyrex
®
disposable culture tubes, screw cap style with white marking spot,
capacity 30 mL
Expand
View Pricing
CLS9944720
Pyrex
®
disposable culture tubes, screw cap style with white marking spot,
capacity 24 mL
Expand
View Pricing
CLS9944716X
Pyrex
®
disposable culture tubes, screw cap style with white marking spot,
capacity 19.0 mL
Expand
View Pricing