Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pullulan Standard
EC Number:
232-945-1
Products
Brand
Supelco (9)
Format
Analyte Suitability
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (9)
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
47053
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 110,000
Expand
View Pricing
43807
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 50,000
Expand
View Pricing
18179
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 6,000
Expand
View Pricing
18579
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 400,000
Expand
View Pricing
42598
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 22,000
Expand
View Pricing
18789
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 800,000
Expand
View Pricing
97873
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 12,000
Expand
View Pricing
53168
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 1,300
Expand
View Pricing
01615
Pullulan Standard,
certified reference material, certified according to DIN, for GPC, 200,000
Expand
View Pricing