
Proteomix® SAX-NP5 (5 µm) HPLC Columns

Products
Product Category
Manufacturer
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing