Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Protein A from
Staphylococcus aureus
Products
Product Category
proteins (4)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Purity
Recombinant Host
Conjugate
Quality Segment
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P6031
Protein A from
Staphylococcus aureus
,
Soluble, essentially salt-free, lyophilized powder, extracellular
Expand
View Pricing
P3838
Protein A from
Staphylococcus aureus
,
Soluble, Cowan Strain, essentially salt-free, lyophilized powder
Expand
View Pricing
P7837
Protein A from
Staphylococcus aureus
,
Soluble, Cowan strain, recombinant, expressed in
E. coli
, aqueous solution, ≥95% (HPLC)
Expand
View Pricing
P7155
Protein A from
Staphylococcus aureus
,
Insoluble, Cowan Strain, cell suspension
Expand
View Pricing