
Prostaglandin A1

Synonym(s):
(13E,15S)-15-Hydroxy-9-oxoprosta-10,13-dien-1-oic acid, PGA1
Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing