
Promethazine for peak identification

Synonym(s):
Promethazine hydrochloride, 10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride
Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing