Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Probumin
®
Bovine Serum Albumin Microbiological Grade
CAS Number:
9048-46-8
Products
Product Category
albumins (3)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Tissue
Quality Segment
Geographic Origin
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
81065N
Probumin
®
Bovine Serum Albumin Microbiological Grade,
≥98%, USA origin, microbiological culture: suitable
Expand
View Pricing
810651
Probumin
®
Bovine Serum Albumin Microbiological Grade,
≥98%, microbiological culture: suitable, 0-5.0% moisture (Karl Fischer), USA origin
Expand
View Pricing
810652
Probumin
®
Bovine Serum Albumin Microbiological Grade,
≥98%, microbiological culture: suitable, 0-5.0% moisture (Karl Fischer), USA origin,
Expand
View Pricing
810653
Probumin
®
Bovine Serum Albumin Microbiological Grade,
≥98%, microbiological culture: suitable, 0-5.0% moisture (Karl Fischer), USA origin
Expand
View Pricing