Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Premium Slide box
Products
Product Category
boxes (20)
Manufacturer
Height (mm)
Color
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
HS15994G
Premium Slide box,
100 Place, cork lining, black
Expand
View Pricing
HS15994E
Premium Slide box,
100 place, cork lining, white plastic
Expand
View Pricing
HS15994M
Premium Slide box,
100 place, foam, blue plastic
Expand
View Pricing
HS15994B
Premium Slide box,
100 Place, cork lining, green
Expand
View Pricing
HS15994A
Premium Slide box,
100 Place, cork lining, blue
Expand
View Pricing
HS15994F
Premium Slide box,
100 place, cork lining, gray plastic
Expand
View Pricing
HS15994D
Premium Slide box,
100 Place, cork lining, yellow
Expand
View Pricing
HS15996C
Premium Slide box,
50 place, cork lining, white plastic
Expand
View Pricing
HS15996A
Premium Slide box,
50 place, cork lining, blue plastic
Expand
View Pricing
HS15989E
Premium Slide box,
25 place, cork lining, white plastic
Expand
View Pricing
HS15994C
Premium Slide box,
100 Place, cork lining, red
Expand
View Pricing
HS15989M
Premium Slide box,
25 place, foam lining, blue plastic
Expand
View Pricing
HS15989B
Premium Slide box,
25 place, cork lining, green plastic
Expand
View Pricing
HS15989R
Premium Slide box,
25 place, foam lining, white plastic
Expand
View Pricing
HS15994R
Premium Slide box,
100 Place, foam lining, white
Expand
View Pricing
HS15989A
Premium Slide box,
25 Place, cork lining, blue
Expand
View Pricing
HS15989P
Premium Slide box,
25 place, foam lining, red plastic
Expand
View Pricing
HS15996B
Premium Slide box,
50 place, cork lining, red plastic
Expand
View Pricing
HS15994P
Premium Slide box,
100 place, foam, red plastic
Expand
View Pricing
HS15989C
Premium Slide box,
25 place, cork lining, red plastic
Expand
View Pricing