Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Premium Plus Slide Box
Products
Product Category
boxes (6)
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
HS120593
Premium Plus Slide Box,
to hold, 100 slide(s), translucent assorted colors acrylonitrile-butadiene-styrene (ABS)
Expand
View Pricing
HS120576
Premium Plus Slide Box,
to hold, 100 slide(s), translucent blue acrylonitrile-butadiene-styrene (ABS)
Expand
View Pricing
HS120577
Premium Plus Slide Box,
to hold, 100 slide(s), translucent orange acrylonitrile-butadiene-styrene (ABS)
Expand
View Pricing
HS120578
Premium Plus Slide Box,
to hold, 100 slide(s), translucent green acrylonitrile-butadiene-styrene (ABS)
Expand
View Pricing
HS120579
Premium Plus Slide Box,
to hold, 100 slide(s), translucent red acrylonitrile-butadiene-styrene (ABS)
Expand
View Pricing
HS120592
Premium Plus Slide Box,
to hold, 100 slide(s), translucent purple acrylonitrile-butadiene-styrene (ABS)
Expand
View Pricing