Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Precision Seal
®
rubber septa
Products
Product Category
septa (22)
Brand
Sigma-Aldrich (4)
Color
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z553964
Precision Seal
®
rubber septa,
white, 14/20 joints
Expand
View Pricing
Z553980
Precision Seal
®
rubber septa,
white, 24/40 joints
Expand
View Pricing
Z553972
Precision Seal
®
rubber septa,
white, 19/22 joints
Expand
View Pricing
Z553999
Precision Seal
®
rubber septa,
white, 29/42 joints
Expand
View Pricing
Z564656
Precision Seal
®
rubber septa,
white, 15/25 joints
Expand
View Pricing
Z553921
Precision Seal
®
rubber septa,
white, 10 mm O.D. glass tubing
Expand
View Pricing
Z554006
Precision Seal
®
rubber septa,
white, mixed set
Expand
View Pricing
Z554073
Precision Seal
®
rubber septa,
red, 14/20 joints
Expand
View Pricing
Z553956
Precision Seal
®
rubber septa,
white, 10/30 joints
Expand
View Pricing
Z553913
Precision Seal
®
rubber septa,
white, 8 mm O.D. glass tubing
Expand
View Pricing
Z553905
Precision Seal
®
rubber septa,
white, 7 mm O.D. glass tubing
Expand
View Pricing
Z553948
Precision Seal
®
rubber septa,
white, 13 mm I.D. glass tubing
Expand
View Pricing
Z554103
Precision Seal
®
rubber septa,
red, 24/40 joints
Expand
View Pricing
Z554081
Precision Seal
®
rubber septa,
red, 19/22 joints
Expand
View Pricing
Z564648
Precision Seal
®
rubber septa,
red, 15/25 joints
Expand
View Pricing
Z554065
Precision Seal
®
rubber septa,
red, 10/30 joints
Expand
View Pricing
Z554111
Precision Seal
®
rubber septa,
red, 29/42 joints
Expand
View Pricing
Z554138
Precision Seal
®
rubber septa,
red, mixed set
Expand
View Pricing
Z554049
Precision Seal
®
rubber septa,
red, 10 mm O.D. glass tubing
Expand
View Pricing
Z554030
Precision Seal
®
rubber septa,
red, 8 mm O.D. glass tubing
Expand
View Pricing
Page 1 of 2
Page 1 of 2