Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium sodium tartrate tetrahydrate
Synonym(s):
Potassium sodium tartrate tetrahydrate, L(+)-Tartaric acid potassium Sodium salt, Rochelle salt, Seignette salt
Linear Formula:
KOCOCH(OH)CH(OH)COONa · 4H
2
O
CAS Number:
6381-59-5
Molecular Weight:
282.22
EC Number:
206-156-8
Products
Product Category
building blocks (1)
salts (1)
Brand
Sigma-Aldrich (5)
Supelco (1)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Optical Isomer
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
1.08087
Potassium sodium tartrate tetrahydrate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
217255
Potassium sodium tartrate tetrahydrate,
ACS reagent, 99%
Expand
View Pricing
S2377
Potassium sodium tartrate tetrahydrate,
ReagentPlus
®
, ≥99%
Expand
View Pricing
32312
Potassium sodium tartrate tetrahydrate,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (perchloric acid titration)
Expand
View Pricing
25508
Potassium sodium tartrate tetrahydrate,
puriss., 99.0-101.0% (calc. on H
2
O free substance), meets analytical specification of Ph. Eur., BP, USP, FC, E 337
Expand
View Pricing
60413
Potassium sodium tartrate tetrahydrate,
BioUltra, ≥99.5% (NT)
Expand
View Pricing