Skip to Content
Merck
Potassium phosphate dibasic trihydrate puriss. p.a., ≥99.0% (T)

Potassium phosphate dibasic trihydrate

Potassium phosphate dibasic trihydrate puriss. p.a., ≥99.0% (T)
Synonym(s):
Dipotassium hydrogen phosphate trihydrate, Dipotassium phosphate, di-Potassium hydrogen phosphate trihydrate
Linear Formula:
K2HPO4 · 3H2O
CAS Number:
Molecular Weight:
228.22
EC Number:

Products

Product Category
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade