Skip to Content
Merck
POPSO hydrate ≥99% (titration)

POPSO hydrate

POPSO hydrate ≥99% (titration)
Synonym(s):
Piperazine-1,4-bis(2-hydroxypropanesulfonic acid) dihydrate, Piperazine-N,N′-bis(2-hydroxypropanesulfonic acid)
Empirical Formula (Hill Notation):
C10H22N2O8S2 · xH2O
CAS Number:
Molecular Weight:
362.42 (anhydrous basis)

Products

Product Category
Formula Weight
pH Value
Physical Form
pK Value
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing