Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(sodium 4-styrenesulfonate) solution
Synonym(s):
PSS, Poly(4-styrenesulfonic acid) sodium salt
Linear Formula:
(C
8
H
7
NaO
3
S)
n
CAS Number:
25704-18-1
Products
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
pH Value
Polymer Composition
Polymer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
561967
Poly(sodium 4-styrenesulfonate) solution,
average M
w
~200,000, 30 wt. % in H
2
O
Expand
View Pricing
527483
Poly(sodium 4-styrenesulfonate) solution,
average M
w
~70,000, 30 wt. % in H
2
O
Expand
View Pricing
527491
Poly(sodium 4-styrenesulfonate) solution,
average M
w
~1,000,000, 25 wt. % in H
2
O
Expand
View Pricing