Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polypyrrole
Synonym(s):
PPy
CAS Number:
30604-81-0
Products
Brand
Sigma-Aldrich (4)
Sustainability
Feature
Melting Point (°C)
Physical Form
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
577030
Polypyrrole,
conductivity 10-50 S/cm (pressed pellet)
Expand
View Pricing
482552
Polypyrrole,
doped, 5 wt % dispersion in H
2
O, conductivity >0.005 S/cm (dried cast film)
Expand
View Pricing
530573
Polypyrrole,
doped, conductivity 30 S/cm (bulk), extent of labeling: 20 wt. % loading, composite with carbon black
Expand
View Pricing
578177
Polypyrrole,
doped, conductivity 0.5-1.5 S/cm (pressed pellet, typical), extent of labeling: ~5 wt. % loading, coated on titanium dioxide
Expand
View Pricing