Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyphosphoric acid
Synonym(s):
Polyphosphoric acid
Linear Formula:
HO[P(OH)(O)O](n)H
CAS Number:
8017-16-1
Products
Product Category
inorganic acids (2)
Brand
Sigma-Aldrich (3)
Sustainability
Boiling Point (°C)
Feature
Melting Point (°C)
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
208213
Polyphosphoric acid,
reagent grade, 115% H
3
PO
4
basis
Expand
View Pricing
398608
Polyphosphoric acid,
~105% H
3
PO
4
basis
Expand
View Pricing
8.07471
Polyphosphoric acid,
for synthesis
Expand
View Pricing