Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly-L-lysine hydrobromide
CAS Number:
25988-63-0
Products
Product Category
amino acids (7)
Brand
Sigma-Aldrich (7)
Sustainability
Color
Feature
Physical Form
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
P2636
Poly-L-lysine hydrobromide,
suitable for cell culture and bioconjugation, Mol wt 30,000-70,000
Expand
View Pricing
P1274
Poly-L-lysine hydrobromide,
suitable for cell culture, Mol wt 70,000-150,000 by viscosity
Expand
View Pricing
P1399
Poly-L-lysine hydrobromide,
suitable for cell culture, Mol wt 150,000-300,000
Expand
View Pricing
P1524
Poly-L-lysine hydrobromide,
suitable for cell culture, Mol wt ≥300,000
Expand
View Pricing
P6516
Poly-L-lysine hydrobromide,
suitable for cell culture, Mol wt ≤15,000 by MALLS
Expand
View Pricing
P7890
Poly-L-lysine hydrobromide,
mol wt 15,000-30,000 by viscosity
Expand
View Pricing
P0879
Poly-L-lysine hydrobromide,
suitable for cell culture, Mol wt 1,000-5,000
Expand
View Pricing