Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly-L-glutamic acid sodium salt
CAS Number:
26247-79-0
Products
Product Category
amino acids (4)
Brand
Sigma-Aldrich (4)
Sustainability
Color
Feature
Physical Form
Application
Optical Isomer
Amino Acid
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P4761
Poly-L-glutamic acid sodium salt,
mol wt 15,000-50,000
Expand
View Pricing
P4636
Poly-L-glutamic acid sodium salt,
suitable for ligand binding assays, Mol wt 3,000-15,000
Expand
View Pricing
P4886
Poly-L-glutamic acid sodium salt,
mol wt 50,000-100,000
Expand
View Pricing
P1818
Poly-L-glutamic acid sodium salt,
mol wt 1,500-5,500 by MALLS
Expand
View Pricing