Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ethylene terephthalate)
Synonym(s):
Poly(ethylene terephthalate)
Linear Formula:
(C
10
H
8
O
4
)
n
CAS Number:
25038-59-9
Products
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Polymer Composition
Polymer Type
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
429252
Poly(ethylene terephthalate),
granular
Expand
View Pricing
1546900
Poly(ethylene terephthalate),
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing