Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polydisperse Particle Standard
Products
Brand
Supelco (4)
Manufacturer
Physical Form
Format
Application
Particle Size (µm)
Reference Material Type
Special Grade
analytical standard (6)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
08718
Polydisperse Particle Standard,
Particle Size Standard, PS315, 10 x 1.0g, 10-100 μm
Expand
View Pricing
94078
Polydisperse Particle Standard,
Particle Size Standard, PS202, 10 x 0.10g, 3-30 μm
Expand
View Pricing
40579
Polydisperse Particle Standard,
Particle Size Standard, PS314, 10 x 0.50g, 10-100 μm
Expand
View Pricing
78456
Polydisperse Particle Standard,
Particle Size Standard, PS226, 10 x 2.5g, 50-350 μm
Expand
View Pricing
57563
Polydisperse Particle Standard,
Particle Size Standard, PS224, 10 x 0.50g, 50-350 μm
Expand
View Pricing
42459
Polydisperse Particle Standard,
Particle Size Standard, PS192, 10 x 0.10g, 1-10 μm
Expand
View Pricing