Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(dimethylsiloxane), hydroxy terminated
Synonym(s):
PDMS
CAS Number:
70131-67-8
Products
Brand
Sigma-Aldrich (6)
Boiling Point (°C)
Physical Form
Quality Segment
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
481939
Poly(dimethylsiloxane), hydroxy terminated,
viscosity ~25 cSt
Expand
View Pricing
481955
Poly(dimethylsiloxane), hydroxy terminated,
viscosity ~65 cSt
Expand
View Pricing
432997
Poly(dimethylsiloxane), hydroxy terminated,
viscosity 18,000-22,000 cSt
Expand
View Pricing
481963
Poly(dimethylsiloxane), hydroxy terminated,
viscosity ~750 cSt
Expand
View Pricing
482005
Poly(dimethylsiloxane), hydroxy terminated,
average M
n
~110,000, viscosity ~50,000 cSt
Expand
View Pricing
482161
Poly(dimethylsiloxane), hydroxy terminated,
viscosity 3,500 cSt
Expand
View Pricing