Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(dimethylsiloxane-
co
-methylphenylsiloxane)
Linear Formula:
[Si(CH
3
)
2
O]
x
[Si(CH
3
)(C
6
H
5
)O]
y
CAS Number:
63148-52-7
Products
Brand
Sigma-Aldrich (3)
Physical Form
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
378488
Poly(dimethylsiloxane-
co
-methylphenylsiloxane),
viscosity 125 cSt
Expand
View Pricing
378445
Poly(dimethylsiloxane-
co
-methylphenylsiloxane),
viscosity 50 cSt
Expand
View Pricing
378461
Poly(dimethylsiloxane-
co
-methylphenylsiloxane),
viscosity 500 cSt
Expand
View Pricing