
p-Mentha-8-thiol-3-one

Synonym(s):
8-Mercaptomenthone, 2-(1-Mercapto-1-methylethyl)-5-methylcyclohexanone
Empirical Formula (Hill Notation):
C10H18OS
Products
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Chemical Composition
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing