Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-iridium alloy, Pt80Ir20
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 29
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF80341321
Platinum-iridium alloy, Pt80Ir20,
0.5m wire, 0.1mm diameter, as drawn
Expand
View Pricing
GF03179774
Platinum-iridium alloy, Pt80Ir20,
0.2m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF56346318
Platinum-iridium alloy, Pt80Ir20,
0.1m wire, 0.34mm diameter, annealed
Expand
View Pricing
GF84004523
Platinum-iridium alloy, Pt80Ir20,
1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF85804229
Platinum-iridium alloy, Pt80Ir20,
0.2m wire, 0.34mm diameter, annealed
Expand
View Pricing
GF34524884
Platinum-iridium alloy, Pt80Ir20,
0.1m wire, 0.50mm diameter, as drawn
Expand
View Pricing
GF31914682
Platinum-iridium alloy, Pt80Ir20,
1m wire, 0.34mm diameter, annealed
Expand
View Pricing
GF44896720
Platinum-iridium alloy, Pt80Ir20,
2m wire, 0.1mm diameter, as drawn
Expand
View Pricing
GF24757960
Platinum-iridium alloy, Pt80Ir20,
1m wire, 0.015mm diameter, stress relieved
Expand
View Pricing
GF97472032
Platinum-iridium alloy, Pt80Ir20,
0.5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF65672635
Platinum-iridium alloy, Pt80Ir20,
0.5m wire, 0.34mm diameter, annealed
Expand
View Pricing
GF18564891
Platinum-iridium alloy, Pt80Ir20,
1m wire, 0.1mm diameter, as drawn
Expand
View Pricing
GF25087877
Platinum-iridium alloy, Pt80Ir20,
1m wire, 0.50mm diameter, as drawn
Expand
View Pricing
GF73759922
Platinum-iridium alloy, Pt80Ir20,
foil, 50 x 50mm, 0.025mm thickness, light tight (lt)
Expand
View Pricing
GF41532410
Platinum-iridium alloy, Pt80Ir20,
foil, 25 x 25mm, 0.025mm thickness, light tight (lt)
Expand
View Pricing
GF74874763
Platinum-iridium alloy, Pt80Ir20,
foil, 25 x 25mm, 0.25mm thickness
Expand
View Pricing
GF11500636
Platinum-iridium alloy, Pt80Ir20,
0.2m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF18492004
Platinum-iridium alloy, Pt80Ir20,
0.1m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF42600620
Platinum-iridium alloy, Pt80Ir20,
0.1m wire, 0.38mm diameter, as drawn
Expand
View Pricing
GF94745807
Platinum-iridium alloy, Pt80Ir20,
5m wire, 0.015mm diameter, stress relieved
Expand
View Pricing
Page 1 of 2
Page 1 of 2