Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-iridium alloy, Pt75Ir25
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF33390637
Platinum-iridium alloy, Pt75Ir25,
25mm rod, 2.9mm diameter, as drawn
Expand
View Pricing
GF51578163
Platinum-iridium alloy, Pt75Ir25,
0.5m wire, 0.1mm diameter, clad with 0.1mm thickness platinum, weight is for the 0.1mm diameter pt75/ir25 core only
Expand
View Pricing
GF64959733
Platinum-iridium alloy, Pt75Ir25,
0.1m wire, 0.1mm diameter, clad with 0.1mm thickness platinum, weight is for the 0.1mm diameter pt75/ir25 core only
Expand
View Pricing
GF47878936
Platinum-iridium alloy, Pt75Ir25,
1m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF48591973
Platinum-iridium alloy, Pt75Ir25,
0.5m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF34863790
Platinum-iridium alloy, Pt75Ir25,
10m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF66900933
Platinum-iridium alloy, Pt75Ir25,
2m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF71181840
Platinum-iridium alloy, Pt75Ir25,
5m wire, 0.05mm diameter, annealed
Expand
View Pricing