Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum/Iridium
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF43243400
Platinum/Iridium,
wire, Pt 70%/Ir 30%, 0.05 mm diameter, length 1 m, temper as drawn
Expand
View Pricing
GF24889562
Platinum/Iridium,
wire, Pt 90%/Ir10%, 0.25 mm diameter, length 0.5 m, temper annealed
Expand
View Pricing
GF14487329
Platinum/Iridium,
rod, Pt 80%/Ir 20%, 1.3 mm diameter, length 200 mm, temper as drawn
Expand
View Pricing
GF36202193
Platinum/Iridium,
wire, Pt 90%/Ir10%, 0.25 mm diameter, length 0.1 m, temper annealed
Expand
View Pricing
GF43900264
Platinum/Iridium,
foil, Pt 80%/Ir 20%, thickness 0.0125 mm, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF72587245
Platinum/Iridium,
wire, 0.25 mm diameter, Pt90%/Ir10%, length 1 m, temper annealed
Expand
View Pricing
GF48336352
Platinum/Iridium,
wire, Pt 70%/Ir 30%, 0.5 mm diameter, length 0.2 m, temper as drawn
Expand
View Pricing
GF36408551
Platinum/Iridium,
wire, Pt85%/Ir 15%, 1.0 mm diameter, length 0.1 m, temper as drawn
Expand
View Pricing
GF93233224
Platinum/Iridium,
wire, Pt90%/Ir10%, 0.25 mm diameter, length 2 m, temper annealed
Expand
View Pricing
GF34668478
Platinum/Iridium,
wire, Pt 70%/Ir 30%, 0.05 mm diameter, length 0.5 m, temper as drawn
Expand
View Pricing
GF50801734
Platinum/Iridium,
wire, Pt 90%/Ir10%, 0.0125 mm diameter, length 1 m, temper as drawn
Expand
View Pricing
GF60131056
Platinum/Iridium,
foil, thickness 0.0125 mm, Pt80%/Ir20%, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF69061877
Platinum/Iridium,
rod, Pt80%/Ir20%, 1.3 mm diameter, length 25 mm, temper as drawn
Expand
View Pricing
GF82818456
Platinum/Iridium,
wire, 0.5 mm diameter, Pt70%/Ir30%, temper as drawn, length 0.1 m
Expand
View Pricing
GF60670436
Platinum/Iridium,
wire, 0.5 mm diameter, Pt70%/Ir30%, length 1 m, temper as drawn
Expand
View Pricing
GF04715557
Platinum/Iridium,
wire, Pt 70%/Ir 30%, 0.5 mm diameter, length 0.5 m, temper as drawn
Expand
View Pricing
GF68764090
Platinum/Iridium,
rod, 1.8 mm diameter, Pt80%/Ir20%, length 10 mm, temper as drawn
Expand
View Pricing
GF25182042
Platinum/Iridium,
wire, Pt 70%/Ir 30%, 0.05 mm diameter, length 2 m, temper as drawn
Expand
View Pricing
GF30137262
Platinum/Iridium,
wire, Pt85%/Ir 15%, 1.0 mm diameter, length 0.2 m, temper as drawn
Expand
View Pricing
GF75203254
Platinum/Iridium,
rod, Pt80%/Ir20%, 1.8 mm diameter, length 20 mm, temper as drawn
Expand
View Pricing
Page 1 of 2
Page 1 of 2