Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips for "OASIS" automated liquid handling system
Products
Product Category
pipette tips (7)
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
AXYOT100R
Pipette tips for "OASIS" automated liquid handling system,
capacity 100 μL, clear
Expand
View Pricing
AXYEVF205WBRS
Pipette tips for "OASIS" automated liquid handling system,
wide bore, racked, filtered, capacity 180 μL, clear
Expand
View Pricing
AXYOTF180RPB
Pipette tips for "OASIS" automated liquid handling system,
with paper band, filtered, clear
Expand
View Pricing
AXYOT200R
Pipette tips for "OASIS" automated liquid handling system,
capacity 200 μL, clear
Expand
View Pricing
AXYEVF205WBRPB
Pipette tips for "OASIS" automated liquid handling system,
with paper band, wide bore, racked, filtered, capacity 180 μL, clear
Expand
View Pricing
AXYOTF50RPB
Pipette tips for "OASIS" automated liquid handling system,
with paper band, filtered, capacity 50 μL, clear
Expand
View Pricing
AXYOTF100RPB
Pipette tips for "OASIS" automated liquid handling system,
with paper band, filtered, capacity 100 μL, clear
Expand
View Pricing