Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Piperitone
Synonym(s):
3-Carvomenthenone, 6-Isopropyl-3-methyl-2-cyclohexen-1-one,
p
-Menth-1-en-3-one
Empirical Formula (Hill Notation):
C
10
H
16
O
CAS Number:
89-81-6
Molecular Weight:
152.23
EC Number:
201-942-7
Products
Brand
Supelco (1)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
analytical standard (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
PHL82627
Piperitone,
phyproof
®
Reference Substance
Expand
View Pricing
79899
Piperitone,
analytical standard
Expand
View Pricing