Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phosphor bronze alloy, Cu94Sn6
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 172
Page 1 of 9
Page 1 of 9
Compare
Product Number
Description
Pricing
GF72579673
Phosphor bronze alloy, Cu94Sn6,
100mm rod, 12.7mm diameter
Expand
View Pricing
GF55922632
Phosphor bronze alloy, Cu94Sn6,
sheet, 150 x 150mm, 3.2mm thickness, hard
Expand
View Pricing
GF44257248
Phosphor bronze alloy, Cu94Sn6,
sheet, 300 x 300mm, 3.2mm thickness, hard
Expand
View Pricing
GF60227434
Phosphor bronze alloy, Cu94Sn6,
foil, 200 x 200mm, 0.4mm thickness, half hard
Expand
View Pricing
GF54639771
Phosphor bronze alloy, Cu94Sn6,
foil, 15mm disks, 0.25mm thickness, hard
Expand
View Pricing
GF73163640
Phosphor bronze alloy, Cu94Sn6,
5m insulated wire, 0.025mm conductor diameter, 0.0025mm insulation thickness, polyimide insulation
Expand
View Pricing
GF15704209
Phosphor bronze alloy, Cu94Sn6,
foil, 300 x 300mm, 0.125mm thickness, hard
Expand
View Pricing
GF97531364
Phosphor bronze alloy, Cu94Sn6,
1000mm rod, 12.7mm diameter
Expand
View Pricing
GF09904338
Phosphor bronze alloy, Cu94Sn6,
500mm rod, 12.7mm diameter
Expand
View Pricing
GF55965921
Phosphor bronze alloy, Cu94Sn6,
foil, 100 x 100mm, 0.076mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF10630932
Phosphor bronze alloy, Cu94Sn6,
100m wire, 0.025mm diameter, hard
Expand
View Pricing
GF16118827
Phosphor bronze alloy, Cu94Sn6,
foil, 5m coil, 20mm coil width, 0.4mm thickness, hard
Expand
View Pricing
GF10574940
Phosphor bronze alloy, Cu94Sn6,
foil, 150 x 150mm, 0.25mm thickness, hard
Expand
View Pricing
GF64067485
Phosphor bronze alloy, Cu94Sn6,
foil, 100 x 100mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF56202500
Phosphor bronze alloy, Cu94Sn6,
foil, 25 x 25mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF16426931
Phosphor bronze alloy, Cu94Sn6,
foil, 100 x 100mm, 0.02mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF57395223
Phosphor bronze alloy, Cu94Sn6,
foil, 8mm disks, 0.005mm thickness, as rolled
Expand
View Pricing
GF45995529
Phosphor bronze alloy, Cu94Sn6,
200mm rod, 25.4mm diameter
Expand
View Pricing
GF24425513
Phosphor bronze alloy, Cu94Sn6,
25m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF34603219
Phosphor bronze alloy, Cu94Sn6,
5m wire, 0.025mm diameter, hard
Expand
View Pricing
Page 1 of 9
Page 1 of 9