Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phosphoglucose Isomerase from baker’s yeast (
S. cerevisiae
)
Synonym(s):
D-Glucose-6-phosphate ketol-isomerase, PGI, Phosphosaccharomutase
CAS Number:
9001-41-6
EC Number:
232-603-1
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Color
Physical Form
Analyte Suitability
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P5381
Phosphoglucose Isomerase from baker’s yeast (
S. cerevisiae
),
Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)
Expand
View Pricing
F2668
Phosphoglucose Isomerase from baker’s yeast (
S. cerevisiae
),
for use with Fructose Assay Kit FA-20
Expand
View Pricing