
Phenylephrine Related Compound C

Synonym(s):
1-(3-Hydroxyphenyl)-2-(methylamino)ethanone hydrochloride, 3′-Hydroxyphenyl-2-methylaminoacetophenone hydrochloride, Phenylephrone hydrochloride
Empirical Formula (Hill Notation):
C9H11NO2 · HCl
CAS Number:
Molecular Weight:
201.65
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2