Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Petri-Seal
™
adhesive sealing film
Products
Product Category
sealing films and foils (5)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Length (m)
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z376922
Petri-Seal
™
adhesive sealing film,
white, roll W × L 12.5 mm × 32.9 m
Expand
View Pricing
Z743564
Petri-Seal
™
adhesive sealing film,
clear, L × W 12.5 in. × 32.9 in.
Expand
View Pricing
Z743563
Petri-Seal
™
adhesive sealing film,
blue, L × W 12.5 in. × 32.9 in.
Expand
View Pricing
Z743566
Petri-Seal
™
adhesive sealing film,
yellow, L × W 12.5 in. × 32.9 in.
Expand
View Pricing
Z743565
Petri-Seal
™
adhesive sealing film,
red, L × W 12.5 in. × 32.9 in.
Expand
View Pricing