
Peonidin 3-glucoside chloride

Synonym(s):
Peonidin 3-O-glucoside chloride, 3-(Glucosyloxy)-4′,5,7-trihydroxy-3′-methoxyflavylium chloride, Glucopeonidin chloride
Empirical Formula (Hill Notation):
C22H23ClO11
CAS Number:
Molecular Weight:
498.86
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing