Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pellicon
®
XL Cassette with Biomax
®
Membrane
Products
Product Category
filters (9)
Brand
Millipore (9)
pH Value
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Membrane Material
MWCO (kDa)
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
PXB010A50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 10 kDa
Expand
View Pricing
PXB030A50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 30 kDa
Expand
View Pricing
PXB050A50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 50 kDa
Expand
View Pricing
PXB100C50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 100 kDa
Expand
View Pricing
PXB300C50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 300 kDa
Expand
View Pricing
PXB005A50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 5 kDa
Expand
View Pricing
PXB500C50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 500 kDa
Expand
View Pricing
PXB01MC50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 1000 kDa
Expand
View Pricing
PXB008A50
Pellicon
®
XL Cassette with Biomax
®
Membrane,
filtration area 50 cm
2
, NMWCO 8 kDa
Expand
View Pricing