
Pellicon® Capsule with Ultracel® Membrane

Synonym(s):
Pellicon® Capsule with Ultracel® Membrane
Products
Product Category
Brand
Product Line
Length (cm)
Length (in)
Volume (ml)
Quality Segment
Housing Material
Membrane Material
Package Size
Sterility
Sterilization Method
Technique
Showing 1-20 of 26
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2