Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pellicon
®
3 Cassette with Ultracel
®
Membrane
Products
Product Category
filters (19)
cassette filters (2)
Brand
Millipore (19)
pH Value
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Membrane Material
MWCO (kDa)
Technique
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
P3C030C00
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 88 cm
2
, NMWCO 30 kDa
Expand
View Pricing
P3C030C01
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.11 m
2
, NMWCO 30 kDa
Expand
View Pricing
P3C030C10
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 1.14 m
2
, NMWCO 30 kDa
Expand
View Pricing
P3C030D00
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 88 cm
2
, NMWCO 30 kDa
Expand
View Pricing
P3C005C10
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
5 kDa, C screen, 1.14 m
2
Expand
View Pricing
P3C010C05
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.57 m
2
, NMWCO 10 kDa
Expand
View Pricing
P3C030D05
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.57 m
2
, NMWCO 30 kDa
Expand
View Pricing
P3C030D01
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.11 m
2
, NMWCO 30 kDa
Expand
View Pricing
P3C030C05
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.57 m
2
, NMWCO 30 kDa
Expand
View Pricing
P3C010C10
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 1.14 m
2
, NMWCO 10 kDa
Expand
View Pricing
P3C010C01
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.11 m
2
, NMWCO 10 kDa
Expand
View Pricing
P3C010C00
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 88 cm
2
, NMWCO 10 kDa
Expand
View Pricing
P3C003C01
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
3 kDa, C screen, 0.11 m
2
Expand
View Pricing
P3C003C05
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.57 m
2
, NMWCO 3 kDa
Expand
View Pricing
P3C003C00
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 88 cm
2
, NMWCO 3 kDa
Expand
View Pricing
P3C030D10
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 1.14 m
2
, NMWCO 30 kDa
Expand
View Pricing
P3C005C00
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 88 cm
2
, NMWCO 5 kDa
Expand
View Pricing
P3C005C05
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 0.57 m
2
, NMWCO 5 kDa
Expand
View Pricing
P3C003C10
Pellicon
®
3 Cassette with Ultracel
®
Membrane,
filtration area 1.14 m
2
, NMWCO 3 kDa
Expand
View Pricing