Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pearl
™
Pipette
Products
Product Category
pipettes (14)
pipettors (14)
Manufacturer
Color
Package Size
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
HS120104
Pearl
™
Pipette,
adjustable, volume 100-1000 μL, blue, single-channel
Expand
View Pricing
HS120106
Pearl
™
Pipette,
adjustable, volume 1000-10000 μL, red, single-channel
Expand
View Pricing
HS120101
Pearl
™
Pipette,
adjustable, volume 10-100 μL, blue, single-channel
Expand
View Pricing
HS120098
Pearl
™
Pipette,
adjustable, volume 0.5-10 μL, light blue, single-channel
Expand
View Pricing
HS120105
Pearl
™
Pipette,
adjustable, volume 500-5000 μL, pink, single-channel
Expand
View Pricing
HS120216
Pearl
™
Pipette,
adjustable, volume 30-300 μL, purple, 8-channel
Expand
View Pricing
HS120102
Pearl
™
Pipette,
adjustable, volume 20-200 μL, yellow, single-channel
Expand
View Pricing
HS120097
Pearl
™
Pipette,
adjustable, volume 0.1-2 μL, green, single-channel
Expand
View Pricing
HS120100
Pearl
™
Pipette,
adjustable, volume 5-50 μL, orange, single-channel
Expand
View Pricing
HS120215
Pearl
™
Pipette,
adjustable, volume 5-50 μL, orange, 8-channel
Expand
View Pricing
HS120218
Pearl
™
Pipette,
adjustable, volume 10-100 μL, blue, 12-channel
Expand
View Pricing
HS120217
Pearl
™
Pipette,
adjustable, volume 5-50 μL, orange, 12-channel
Expand
View Pricing
HS120099
Pearl
™
Pipette,
adjustable, volume 2-20 μL, light purple, single-channel
Expand
View Pricing
HS120103
Pearl
™
Pipette,
adjustable, volume 30-300 μL, purple, single-channel
Expand
View Pricing