Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Papain from papaya latex
Synonym(s):
Papainase
CAS Number:
9001-73-4
EC Number:
232-627-2
Products
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Quality Segment
Application
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P4762
Papain from papaya latex,
lyophilized powder, ≥10 units/mg protein
Expand
View Pricing
P3125
Papain from papaya latex,
buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein
Expand
View Pricing
P5306
Papain from papaya latex,
lyophilized powder, aseptically filled
Expand
View Pricing
P3375
Papain from papaya latex,
crude powder, 1.5-10 units/mg solid
Expand
View Pricing