Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium on activated charcoal
Synonym(s):
Pd/C
Linear Formula:
Pd
Products
Brand
Sigma-Aldrich (4)
Sustainability
Feature
Physical Form
Element
Reaction Type
Reagent Type
Core
Special Grade
Puriss (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
75990
Palladium on activated charcoal,
10% Pd basis
Expand
View Pricing
75993
Palladium on activated charcoal,
moistened with water, 10% Pd basis (based on dry substance)
Expand
View Pricing
75992
Palladium on activated charcoal,
5% Pd basis
Expand
View Pricing
75991
Palladium on activated charcoal,
moistened with water, 5% Pd basis (based on dry substance)
Expand
View Pricing