Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium
Synonym(s):
Palladium
Linear Formula:
Pd
CAS Number:
7440-05-3
Molecular Weight:
106.42
Products
Brand
Sigma-Aldrich (13)
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Element
Quality Segment
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
203939
Palladium,
powder, 99.995% trace metals basis
Expand
View Pricing
326666
Palladium,
powder, <1 μm, ≥99.9% trace metals basis
Expand
View Pricing
686468
Palladium,
nanopowder, <25 nm particle size (TEM), ≥99.5%
Expand
View Pricing
203998
Palladium,
powder or granules, 99.99% trace metals basis
Expand
View Pricing
267120
Palladium,
foil, thickness 0.025 mm, 99.9% trace metals basis
Expand
View Pricing
267082
Palladium,
sponge, 99.9% trace metals basis
Expand
View Pricing
464651
Palladium,
powder, <75 μm, 99.9% trace metals basis
Expand
View Pricing
348694
Palladium,
wire, diam. 0.5 mm, 99.9% trace metals basis
Expand
View Pricing
348678
Palladium,
foil, thickness 1.0 mm, 99.9% trace metals basis
Expand
View Pricing
267112
Palladium,
wire, diam. 1.0 mm, 99.9% trace metals basis
Expand
View Pricing
287474
Palladium,
foil, thickness 0.5 mm, 99.9% trace metals basis
Expand
View Pricing
348643
Palladium,
foil, thickness 0.25 mm, 99.98% trace metals basis
Expand
View Pricing
GF39094904
Palladium,
tube, 50mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Expand
View Pricing
373206
Palladium,
evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Expand
View Pricing
GF19845120
Palladium,
rod, 50mm, diameter 4.0mm, 99.95%
Expand
View Pricing
GF81435146
Palladium,
rod, 100mm, diameter 2.0mm, 99.95%
Expand
View Pricing
GF35373551
Palladium,
foil, 10x10mm, thickness 0.50mm, as rolled, 99.99+%
Expand
View Pricing
GF73152648
Palladium,
rod, 10mm, diameter 6.0mm, 99.95%
Expand
View Pricing
GF17478445
Palladium,
foil, 25x25mm, thickness 0.50mm, as rolled, 99.99+%
Expand
View Pricing
GF99541985
Palladium,
tube, 200mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Expand
View Pricing
Page 1 of 11
Page 1 of 11