Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Oxygen-
18
O
2
Linear Formula:
18
O
2
CAS Number:
32767-18-3
Molecular Weight:
36.00
Products
Product Category
gases (6)
stable isotopes (6)
Brand
Sigma-Aldrich (6)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
490474
Oxygen-
18
O
2
,
97 atom %
18
O
Expand
View Pricing
602892
Oxygen-
18
O
2
,
99 atom %
18
O, 99% (CP)
Expand
View Pricing
602876
Oxygen-
18
O
2
,
90 atom %
18
O
Expand
View Pricing
602868
Oxygen-
18
O
2
,
50 atom %
18
O (random)
Expand
View Pricing
602914
Oxygen-
18
O
2
,
10 atom %
18
O (random)
Expand
View Pricing
602922
Oxygen-
18
O
2
,
25 atom %
18
O (random)
Expand
View Pricing