Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Oxygen-
17
O
2
Linear Formula:
17
O
2
CAS Number:
67321-81-7
Molecular Weight:
34.00
Products
Product Category
gases (10)
stable isotopes (10)
building blocks (1)
Brand
Sigma-Aldrich (10)
Formula Weight
Physical Form
Isotope
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
602841
Oxygen-
17
O
2
,
90 atom %
17
O
Expand
View Pricing
602779
Oxygen-
17
O
2
,
70 atom %
17
O
Expand
View Pricing
602833
Oxygen-
17
O
2
,
85 atom %
17
O
Expand
View Pricing
602825
Oxygen-
17
O
2
,
80 atom %
17
O
Expand
View Pricing
602787
Oxygen-
17
O
2
,
10 atom %
17
O
Expand
View Pricing
602760
Oxygen-
17
O
2
,
20 atom %
17
O
Expand
View Pricing
602795
Oxygen-
17
O
2
,
40 atom %
17
O
Expand
View Pricing
602817
Oxygen-
17
O
2
,
45 atom %
17
O
Expand
View Pricing
602809
Oxygen-
17
O
2
,
60 atom %
17
O
Expand
View Pricing
609684
Oxygen-
17
O
2
,
50 atom %
17
O
Expand
View Pricing