Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Optional heads for Corning
®
LSE
™
vortex mixers
Products
Manufacturer
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS480101
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head for 24 x 1.5/2.0 mL tubes, 24 x 0.5 mL tubes and 32 x 0.2 mL tubes (or 4 tube strips), 1/cs
Expand
View Pricing
CLS480105
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head for 12 x 1.5/2.0 mL tubes (held horizontally), 1/cs
Expand
View Pricing
CLS480100
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head, 3 inch, for processing larger numbers of tubes, 1/cs
Expand
View Pricing
CLS480104
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head for 6 x 50 mL tubes, 1/cs
Expand
View Pricing
CLS480102
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head for 1 microplate, 64 x 0.2 mL tubes, or 8 x 0.2 mL tube strips, 1/cs
Expand
View Pricing
CLS480107
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head for 2 x 50 mL tubes (held horizontally), 1/cs
Expand
View Pricing
CLS480106
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head for 4 x 15 mL tubes (held horizontally), 1/cs
Expand
View Pricing
CLS480103
Optional heads for Corning
®
LSE
™
vortex mixers,
optional head for 8 x 15 mL and 8 x 12/13 mm diameter tubes, 1/cs
Expand
View Pricing